Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Diethylamino)Sulfur Trifluoride

Catalog No. 2352535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-525-35G 5 g
23-525-31G 1 g
23-525-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 23-525-35G Supplier Sigma Aldrich Supplier No. 2352535G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fluorinating agent: reaction with alcohols and carbonyl compounds, ReviewReview on nucleophilic fluorination.Catalyst for Friedel-Crafts allylation using tertiary cyclopropyl silyl ethers and the rearrangement of homoallylic alcohols to unsaturated aldehydes.Early introduction of a fluoromethyl group stabilizes the epoxide during further manipulations in the synthesis of 26-fluoro-epothilone.Fluorinating agent for a variety of compounds, including thioethers, alkenols, and cyanohydrins.Reagent for gem difluorination of ketopipecolinic acids.

Specifications

CAS 38078-09-0
Density 1.22 g/mL at 25°C
Boiling Point 30°C to 32°C (3 mmHg, lit.)
Molecular Formula C4H10F3NS
Linear Formula (C2H5)2NSF3
MDL Number MFCD00000363
Quantity 5 g
Synonym DAST; Diethylaminosulfur trifluoride
Molecular Weight (g/mol) 161.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.