Learn More
AdipoGen Diethylaminocoumarin-3 COOH im
SDP
Supplier:  AdipoGen CDXD0071G001
Encompass_Preferred
Chemical. CAS 261943-47-9. Formula C17H17N3O3. MW 311.34. Synthetic. fluorescent reagent for use in circular dichroic exciton method for derivatisation of hydroxyl-groups under mild conditions. The absorption at 406 nm is futher in the red than for reagents like naphthoylimidazole. The reagent offers high sensitivity through a high molar extinction coefficient.
Catalog No. 50-256-1264