Learn More
Medchemexpress LLC Diethylaminoethyl dextran | 9015-73-0 | MFCD00081561 | >=95.0% | 40,000 Da | 250 MG
SDP
Catalog No. 5000283345
Encompass_Preferred
DEAE-dextran is a cationic diethylaminoethyl-substituted dextran polymer (CAS 9015-73-0) with an average molecular weight of about 40,000 Da. Supplied as a dry reagent for life-science research, it is commonly used to facilitate nucleic acid delivery, enhance precipitation of polyanions, and serve as a cationic carrier in molecular biology workflows.
  • Cationic polymer used for transfection and nucleic acid delivery.
  • Average molecular weight approximately 40,000 Da.
  • Water-soluble powder for solution preparation.
  • Facilitates complexation and precipitation of polyanionic biomolecules.
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-002-83345 Supplier Medchemexpress LLC Supplier No. HYW134327250MG
Add to Cart
Add to Cart
DEAE-dextran is a cationic diethylaminoethyl-substituted dextran polymer (CAS 9015-73-0) with an average molecular weight of about 40,000 Da. Supplied as a dry reagent for life-science research, it is commonly used to facilitate nucleic acid delivery, enhance precipitation of polyanions, and serve as a cationic carrier in molecular biology workflows.
  • Cationic polymer used for transfection and nucleic acid delivery.
  • Average molecular weight approximately 40,000 Da.
  • Water-soluble powder for solution preparation.
  • Facilitates complexation and precipitation of polyanionic biomolecules.
  • Supplied in small research pack sizes for laboratory use.