Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylbenzene (mixture) 98.0+%, TCI America™
SDP

Catalog No. D337825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D337825ML 25 mL
D3378500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D337825ML Supplier TCI America Supplier No. D337825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25340-17-4
Molecular Formula C10H14
MDL Number MFCD00792900

Specifications

Melting Point -31°C
Color Colorless
Boiling Point 184°C
Quantity 25 mL
UN Number 2049
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylbenzene (mixture)
EINECSNumber (3)-0013&(3)-0060
RTECSNumber CZ5600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.