Learn More
Medchemexpress LLC Diethylcarbamazine | 90-89-1 | 99.8% | 199.29 | 1 ML
SDP
Catalog No. 5000438041
Encompass_Preferred
Diethylcarbamazine is an orally active microfilaricidal agent, effective in onchocerciasis and lymphatic filariasis. It reduces eosinophil trafficking to lung tissue, exhibiting anti-allergic effects. The compound ameliorates insulin resistance by lowering serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides. It also enhances reactive oxygen intermediate expression by polymorphonuclear neutrophils, increases lymphocyte proliferation, and inhibits actinomycetoma lesion development. Research applications include bronchial asthma, insulin resistance, and infection.
  • Orally active microfilaricidal
  • Anti-allergic properties
  • Ameliorates insulin resistance
  • Modulates immune response
  • Inhibits actinomycetoma lesion development
  • Relevant for bronchial asthma, insulin resistance, and infection research
Catalog No. 50-004-38041 Supplier Medchemexpress LLC Supplier No. HY1264210MM/1ML
Diethylcarbamazine is an orally active microfilaricidal agent, effective in onchocerciasis and lymphatic filariasis. It reduces eosinophil trafficking to lung tissue, exhibiting anti-allergic effects. The compound ameliorates insulin resistance by lowering serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides. It also enhances reactive oxygen intermediate expression by polymorphonuclear neutrophils, increases lymphocyte proliferation, and inhibits actinomycetoma lesion development. Research applications include bronchial asthma, insulin resistance, and infection.
  • Orally active microfilaricidal
  • Anti-allergic properties
  • Ameliorates insulin resistance
  • Modulates immune response
  • Inhibits actinomycetoma lesion development
  • Relevant for bronchial asthma, insulin resistance, and infection research