Learn More
Medchemexpress LLC Diethylcarbamazine | 90-89-1 | 99.8% | 199.29 | 500 MG
SDP
Catalog No. 5000435787
Encompass_Preferred
Diethylcarbamazine is an orally active microfilaricidal agent, effective in treating onchocerciasis and lymphatic filariasis. It exhibits anti-allergic effects by reducing eosinophil trafficking to lung tissue and modulates several biological pathways, including decreasing serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides, thereby ameliorating insulin resistance.
  • Reduces eosinophil trafficking to lung tissue
  • Exerts anti-allergic effects
  • Decreases serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides
  • Ameliorates insulin resistance
  • Enhances reactive oxygen intermediate expression by polymorphonuclear neutrophils
  • Increases lymphocyte proliferation
  • Inhibits actinomycetoma lesion development
  • Suitable for research in bronchial asthma, insulin resistance, and infection
Catalog No. 50-004-35787 Supplier Medchemexpress LLC Supplier No. HY12642A500MG
Add to Cart
Add to Cart
Diethylcarbamazine is an orally active microfilaricidal agent, effective in treating onchocerciasis and lymphatic filariasis. It exhibits anti-allergic effects by reducing eosinophil trafficking to lung tissue and modulates several biological pathways, including decreasing serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides, thereby ameliorating insulin resistance.
  • Reduces eosinophil trafficking to lung tissue
  • Exerts anti-allergic effects
  • Decreases serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides
  • Ameliorates insulin resistance
  • Enhances reactive oxygen intermediate expression by polymorphonuclear neutrophils
  • Increases lymphocyte proliferation
  • Inhibits actinomycetoma lesion development
  • Suitable for research in bronchial asthma, insulin resistance, and infection