Learn More
Medchemexpress LLC Diethylcarbamazine citrate | 1642-54-2 | 391.42 | 500 MG
SDP
Catalog No. 5000438060
Encompass_Preferred
Diethylcarbamazine citrate (Standard) is the analytical standard of Diethylcarbamazine (citrate), intended for research and analytical applications. It is an orally active anthropoidal compound that inhibits arachidonic acid metabolism of filaria microfilaria, exhibiting anti-inflammatory and antiparasitic activity. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Orally active anthropoidal compound
  • Inhibitor of arachidonic acid metabolism of filaria microfilaria
  • Exhibits anti-inflammatory and antiparasitic activity
  • Used as an analytical standard and reference standard assay
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-38060 Supplier Medchemexpress LLC Supplier No. HY12642500MG
Diethylcarbamazine citrate (Standard) is the analytical standard of Diethylcarbamazine (citrate), intended for research and analytical applications. It is an orally active anthropoidal compound that inhibits arachidonic acid metabolism of filaria microfilaria, exhibiting anti-inflammatory and antiparasitic activity. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Orally active anthropoidal compound
  • Inhibitor of arachidonic acid metabolism of filaria microfilaria
  • Exhibits anti-inflammatory and antiparasitic activity
  • Used as an analytical standard and reference standard assay
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS