Learn More
TARGETMOL CHEMICALS INC Diethylcarbamazine citrate 1G
SDP
Catalog No. 502879589
Encompass_Preferred
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Diethylcarbamazine citrate (Caritrol) is an anthelmintic used primarily as the citrate in the treatment of filariasis, particular infestations with Wucheria bancrofti or Loa loa. It is an inhibitor of arachidonic acid metabolism in filarial microfilaria. Purity 99.97%
Catalog No. 50-287-9589 Supplier TARGETMOL CHEMICALS INC Supplier No. T14201G
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Diethylcarbamazine citrate (Caritrol) is an anthelmintic used primarily as the citrate in the treatment of filariasis, particular infestations with Wucheria bancrofti or Loa loa. It is an inhibitor of arachidonic acid metabolism in filarial microfilaria. Purity 99.97%