Learn More
Selleck Chemical LLC Diethylcarbamazine citrate S4564-100mg
SDP
Catalog No. 503108943
Encompass_Preferred
Diethylcarbamazine citrate is an inhibitor of arachidonic acid metabolism in filarial microfilaria and is highly specific for several parasites and does not contain any toxic metallic elements
Catalog No. 50-310-8943 Supplier Selleck Chemical LLC Supplier No. S4564100MG
Diethylcarbamazine citrate is an inhibitor of arachidonic acid metabolism in filarial microfilaria and is highly specific for several parasites and does not contain any toxic metallic elements