Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylcarbamoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D048225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D048225G 25 g
D0482500G 500 g
D0482100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D048225G Supplier TCI America Supplier No. D048225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-10-8
Molecular Formula C5H10ClNO
Molecular Weight (g/mol) 135.591
MDL Number MFCD00000636
InChI Key OFCCYDUUBNUJIB-UHFFFAOYSA-N
Synonym diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride
PubChem CID 6916
IUPAC Name N,N-diethylcarbamoyl chloride
SMILES CCN(CC)C(=O)Cl

Specifications

Boiling Point 195°C
Color Yellow
Quantity 25 g
Formula Weight 135.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylcarbamoyl Chloride
EINECSNumber (2)-2859
RTECSNumber FD4025000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.