Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylcyanamide 98.0+%, TCI America™
SDP

Catalog No. D49655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49655G 5 g
D49651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49655G Supplier TCI America Supplier No. D49655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 617-83-4
Molecular Formula C5H10N2
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001769
InChI Key ZZTSQZQUWBFTAT-UHFFFAOYSA-N
Synonym n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
PubChem CID 12050
IUPAC Name cyanodiethylamine
SMILES CCN(CC)C#N

Specifications

Color Yellow
Boiling Point 188°C
Quantity 5 g
UN Number 3276
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylcyanamide
RTECSNumber GS6300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.