Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyldimethylammonium Hydroxide Solution

Catalog No. 3681350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-813-50ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 36-813-50ML Supplier Sigma Aldrich Supplier No. 3681350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyldimethylammonium hydroxide can be used:As a sole structure-directing agent in the synthesis of zeolite UZM-8 which is used as a catalyst in isopropylation of benzene to form cumene.As an organic additive in the synthesis zeolite Y nanoparticles by hydrothermal method.To synthesize layered silicates PLS-3 and PLS-4, which is used as a precursor for topotactic zeolites.To synthesize layered uranyl diphosphonate compounds from uranyl nitrate and 1,4-benzenebisphosphonic acid by hydrothermal method.

Specifications

CAS 1067-71-6
Density 1.01 g/mL at 25°C
Molecular Formula C7H15O4P
Linear Formula CH3COCH2P(O)(OC2H5)2
MDL Number MFCD00044728
Quantity 25 g
Molecular Weight (g/mol) 194.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.