Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyldithiocarbamic Acid, Diethylammonium Salt 98%, Thermo Scientific™

Catalog No. AC407281000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC407281000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407281000 Supplier Thermo Scientific Chemicals Supplier No. 407281000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1518-58-7
Quantity 100 g
Assay Percent Range 98%
Molecular Formula C5H11NS2·C4H11N
Linear Formula (C2H5)2NCS2·NH2(C2H5)2
MDL Number MFCD00013146
Beilstein 04, IV, 389
Formula Weight 222.42
Percent Purity ≥97.5%
Infrared Spectrum Authentic
Chemical Name or Material Diethyldithiocarbamic acid, diethylammonium salt, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 216-175-3
RTECSNumber EZ5425000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.