Learn More
Medchemexpress LLC Diethylene glycol | 111-46-6 | MFCD00002882 | 99.9% | 5 KG
SDP
Catalog No. 5000411798
Encompass_Preferred
Diethylene glycol (2,2'-Oxybis(ethan-1-ol)) exhibits excellent solubility and reactivity in certain chemical reactions, potentially accelerating the activation of some reactants. It is also widely used in the production of pigments, resins, and other chemicals.
  • Excellent solubility
  • Reactivity in certain chemical reactions
  • Potentially accelerates activation of some reactants
  • Widely used in the production of pigments
  • Widely used in the production of resins
  • Widely used in the production of other chemicals
Catalog No. 50-004-11798 Supplier Medchemexpress LLC Supplier No. HYB19525KG
Diethylene glycol (2,2'-Oxybis(ethan-1-ol)) exhibits excellent solubility and reactivity in certain chemical reactions, potentially accelerating the activation of some reactants. It is also widely used in the production of pigments, resins, and other chemicals.
  • Excellent solubility
  • Reactivity in certain chemical reactions
  • Potentially accelerates activation of some reactants
  • Widely used in the production of pigments
  • Widely used in the production of resins
  • Widely used in the production of other chemicals