Learn More
Medchemexpress LLC Diethylene glycol bis(p-toluenesulfonate) | 7460-82-4 | 99.8% | 414.49 | 100 G
SDP
Catalog No. 5000366316
Encompass_Preferred
Diethylene glycol bis(p-toluenesulfonate) is a PEG-based linker utilized in the synthesis of targeted protein degraders. These degraders are composed of two distinct ligands joined by a linker, where one ligand binds to an E3 ubiquitin ligase and the other binds to the target protein. This mechanism allows for the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system.
  • Functions as a PEG-based linker for molecular synthesis.
  • Enables targeted protein degradation.
  • Utilizes the ubiquitin-proteasome system.
Catalog No. 50-003-66316 Supplier Medchemexpress LLC Supplier No. HYW010948100G
Diethylene glycol bis(p-toluenesulfonate) is a PEG-based linker utilized in the synthesis of targeted protein degraders. These degraders are composed of two distinct ligands joined by a linker, where one ligand binds to an E3 ubiquitin ligase and the other binds to the target protein. This mechanism allows for the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system.
  • Functions as a PEG-based linker for molecular synthesis.
  • Enables targeted protein degradation.
  • Utilizes the ubiquitin-proteasome system.