Learn More
Medchemexpress LLC Diethylene glycol diethyl ether | 112-36-7 | MFCD00009254 | 500g
SDP
Catalog No. 5000266331
Encompass_Preferred
Diethylene glycol diethyl ether is an electrolyte solvent for a non-aqueous lithium-air cell Diethylene glycol diethyl ether can facilitate the reversible reduction and oxidation processes at the porous carbon electrode without a catalyst[1]
Catalog No. 50-002-66331 Supplier Medchemexpress LLC Supplier No. HYY1810500G
Diethylene glycol diethyl ether is an electrolyte solvent for a non-aqueous lithium-air cell Diethylene glycol diethyl ether can facilitate the reversible reduction and oxidation processes at the porous carbon electrode without a catalyst[1]