Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol dimethyl ether, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020548 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0548 1 mL
11-101-9160 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0548 Supplier MilliporeSigma Supelco Supplier No. 041431MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethylene glycol dimethyl ether is a combustible liquid, which can be used in the synthesis of films, chemically similar to polyethylene glycol on low-density polyethylene and Si(100) surfaces via plasma polymerization and deposition.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 370°F
CAS 111-96-6
Vapor Pressure 3 mmHg (20°C)
Vapor Density 4.6 (vs. air)
Density 0.944 g/mL at 20°C; 0.939 g/mL (at 25°C)
Flash Point 57°C
Linear Formula (CH3 OCH2 CH2)2 O
MDL Number MFCD00008503
Melting Point -64°C
Boiling Point 162°C
Synonym 2-Methoxyethyl ether; Bis(2-methoxyethyl) ether; Dimethyldiglycol; ′Diglyme’
Type Analytical Standard
Quantity 1 mL
Formula Weight 134.17
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.