Learn More
Medchemexpress LLC Diethylene glycol mo 1kg
SDP
Catalog No. 5000206434
Encompass_Preferred
Diethylene glycol monobutyl ether acetate is an orally available high molecular weight glycol ether solvent that can be used as a coalescing agent for polyvinyl acetate (latex) coatings the acute oral LD in mice and rats is about 7 g/kg[1]
Catalog No. 50-002-06434 Supplier Medchemexpress LLC Supplier No. HYW0994371KG
Diethylene glycol monobutyl ether acetate is an orally available high molecular weight glycol ether solvent that can be used as a coalescing agent for polyvinyl acetate (latex) coatings the acute oral LD in mice and rats is about 7 g/kg[1]