Learn More
Medchemexpress LLC Diethylene glycol mo 500g
SDP
Catalog No. 5000206853
Encompass_Preferred
Diethylene glycol monobutyl ether acetate is an orally available high molecular weight glycol ether solvent that can be used as a coalescing agent for polyvinyl acetate (latex) coatings the acute oral LD in mice and rats is about 7 g/kg[1]
Catalog No. 50-002-06853 Supplier Medchemexpress LLC Supplier No. HYW099437500G
Diethylene glycol monobutyl ether acetate is an orally available high molecular weight glycol ether solvent that can be used as a coalescing agent for polyvinyl acetate (latex) coatings the acute oral LD in mice and rats is about 7 g/kg[1]