Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monoethyl Ether 99.0+%, TCI America™
SDP

Catalog No. E0048500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0048500G 500 g
E004825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0048500G Supplier TCI America Supplier No. E0048500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 111-90-0 | C6H14O3 | 134.175 g/mol

Chemical Identifiers

Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym Carbitol, 2-(2-Ethoxyethoxy)ethanol
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

CAS 111-90-0
Boiling Point 196°C
Color Colorless
Physical Form Clear Liquid at 20°C
Quantity 500 g
Formula Weight 134.18
Percent Purity ≥99.0% (GC)
Chemical Name or Material Diethylene Glycol Monoethyl Ether
EINECSNumber (2)-0422
RTECSNumber KK8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.