Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethylene glycol monohexyl ether | 112-59-4 | MFCD00010703 | >=95.0% | 190.28 g/mol | C10H22O3 | 100 G
SDP

Catalog No. 5000283393
Encompass_Preferred

Diethylene glycol monohexyl ether is a liquid reagent used as a solvent and additive in biochemical assays and research applications. It functions as a polar, high-boiling solvent suitable for formulations, extraction, and analytical protocols; supplied in laboratory-scale packages for research use.

  • Used as a solvent in biochemical assays.
  • Colorless to light yellow liquid.
  • CAS number 112-59-4 for unambiguous identification.
  • Available in 100 G, 500 G, and 1 KG packages.
  • Chemical formula C10H22O3; molecular weight 190.28 g/mol.
  • Typical purity available ≥95.0%, varies by supplier.

Catalog No. 50-002-83393 Supplier Medchemexpress LLC Supplier No. HYW133950100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethylene glycol monohexyl ether is a liquid reagent used as a solvent and additive in biochemical assays and research applications. It functions as a polar, high-boiling solvent suitable for formulations, extraction, and analytical protocols; supplied in laboratory-scale packages for research use.

  • Used as a solvent in biochemical assays.
  • Colorless to light yellow liquid.
  • CAS number 112-59-4 for unambiguous identification.
  • Available in 100 G, 500 G, and 1 KG packages.
  • Chemical formula C10H22O3; molecular weight 190.28 g/mol.
  • Typical purity available ≥95.0%, varies by supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.