Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monophenyl Ether, MP Biomedicals

Catalog No. ICN227401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227401 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227401 Supplier MP Biomedicals, Inc Supplier No. 227401

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104-68-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00045989
InChI Key ZUAURMBNZUCEAF-UHFFFAOYSA-N
Synonym 2-2-phenoxyethoxy ethanol, phenyl carbitol, diethylene glycol monophenyl ether, phenoxydiglycol, ethanol, 2-2-phenoxyethoxy, fenylkarbitol, diethylene glycolphenyl ether, 2-2-phenoxyethoxy ethan-1-ol, diethylene glycol phenyl ether, fenylkarbitol czech
PubChem CID 7715
IUPAC Name 2-(2-phenoxyethoxy)ethan-1-ol
SMILES OCCOCCOC1=CC=CC=C1

Specifications

Quantity 100 g
Chemical Name or Material Diethylene Glycol Monophenyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.