Learn More
Medchemexpress LLC Diethylene glycol (Standard) | 111-46-6 | 99.9% | 500 MG
SDP
Catalog No. 5000411246
Encompass_Preferred
Diethylene glycol (Standard) is the analytical standard of Diethylene glycol, intended for research and analytical applications. It has excellent solubility and reactivity in certain chemical reactions, helping to accelerate the activation of certain reactants. It is also widely used in the production process of pigments, resins, and other chemicals. It has wide applications in chemical research and industrial production, though it has no direct medical application.
  • Analytical standard for Diethylene glycol.
  • Excellent solubility and reactivity in certain chemical reactions.
  • Can help accelerate the activation of certain reactants.
  • Widely used in the production process of pigments, resins and other chemicals.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-11246 Supplier Medchemexpress LLC Supplier No. HYB1952R500MG
Diethylene glycol (Standard) is the analytical standard of Diethylene glycol, intended for research and analytical applications. It has excellent solubility and reactivity in certain chemical reactions, helping to accelerate the activation of certain reactants. It is also widely used in the production process of pigments, resins, and other chemicals. It has wide applications in chemical research and industrial production, though it has no direct medical application.
  • Analytical standard for Diethylene glycol.
  • Excellent solubility and reactivity in certain chemical reactions.
  • Can help accelerate the activation of certain reactants.
  • Widely used in the production process of pigments, resins and other chemicals.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.