Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethylene glycol (standard) | 111-46-6 | MFCD00002882 | 99.9% | 50 MG
SDP

Catalog No. 5000412189
Encompass_Preferred

Diethylene glycol (standard) | 111-46-6 | MFCD00002882 | 99.9% | 50 MG

Catalog No. 50-004-12189 Supplier Medchemexpress LLC Supplier No. HYB1952R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethylene glycol (standard) | 111-46-6 | MFCD00002882 | 99.9% | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.