Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylenetriamine

Catalog No. D9385625L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
10 g
100 mL
2.5 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9385625L 2.5 L
D9385618L 18 L
D938565ML 5 mL
D93856100ML 100 mL
D938561L 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. D9385625L Supplier Sigma Aldrich Supplier No. D938562.5L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylenetriamine

Specifications

CAS 111-40-0
Melting Point -35°C
Density 0.955 g/mL at 25°C
Boiling Point 199°C to 209°C
Molecular Formula C4H13N3
Refractive Index n20/D 1.484
Linear Formula (NH2CH2CH2)2NH
MDL Number MFCD00008171
Quantity 2.5 L
Synonym 2,2'-Diaminodiethylamine; 2,2'-Iminodiethylamine; Bis(2-aminoethyl)amine
Molecular Weight (g/mol) 103.17 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.