Learn More
Strem, An Ascensus Company CAS# 111-40-0. 500g. Diethylenetriamine, min. 95% DIEN. MFCD00008171
SDP
Catalog No. 5090108994
Encompass
CAS# 111-40-0. 500g. Diethylenetriamine, min. 95% DIEN. MFCD00008171. Molecular Weight: 103.17. Molecular Formula: NH2CH2CH2NHCH2CH2NH2. Color/form: colorless liq. Strem# 07-0400. http://www.strem.com/catalog/v/07-0400/
Catalog No. 50-901-08994 Supplier Strem, An Ascensus Company Supplier No. 070400500G
CAS# 111-40-0. 500g. Diethylenetriamine, min. 95% DIEN. MFCD00008171. Molecular Weight: 103.17. Molecular Formula: NH2CH2CH2NHCH2CH2NH2. Color/form: colorless liq. Strem# 07-0400. http://www.strem.com/catalog/v/07-0400/