Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriamine-N,N,N',N″l,N″l-Pentaacetic Acid, MP Biomedicals™

Catalog No. MP215147825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215147825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215147825 Supplier MP Biomedicals, Inc Supplier No. 0215147825
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a chelating agent. Diethylenetriamine-N,N,N',N'l,N'l-Pentaacetic Acid, MP Biomedicals may be used to synthesize chitosan (CH)-gadolinium (Gd) diethylenetriaminepentaacetic acid (DTPA) conjugates.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-43-6
Molecular Formula C14H23N3O10
Molecular Weight (g/mol) 393.349
InChI Key QPCDCPDFJACHGM-UHFFFAOYSA-N
Synonym pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
PubChem CID 3053
ChEBI CHEBI:35739
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Melting Point 219°C to 220°C (literature)
Density 1.770g/mL (Lit.)
pH 2.27 (literature)
Quantity 25 g
Solubility Information Soluble in 1M hydrochloric acid (50mg/mL) and water.
Formula Weight 393.35
Percent Purity ∼90%
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.