Learn More
Strem, An Ascensus Company CAS# 67-43-6. 50g. Diethylenetriaminepentaacetic acid, 98.5% DTPA (USP). MFCD00004289
SDP
Catalog No. 5081466
Encompass
CAS# 67-43-6. 50g. Diethylenetriaminepentaacetic acid, 98.5% DTPA (USP). MFCD00004289. Molecular Weight: 393.35. Molecular Formula: (HO2CCH2)2N(CH2)2N(CH2CO2H)(CH2)2N(CH2CO2H)2. Color/form: white xtl. Strem# 07-0398. http://www.strem.com/catalog/v/07-0398
Catalog No. 50-814-66 Supplier Strem, An Ascensus Company Supplier No. 07039850G
CAS# 67-43-6. 50g. Diethylenetriaminepentaacetic acid, 98.5% DTPA (USP). MFCD00004289. Molecular Weight: 393.35. Molecular Formula: (HO2CCH2)2N(CH2)2N(CH2CO2H)(CH2)2N(CH2CO2H)2. Color/form: white xtl. Strem# 07-0398. http://www.strem.com/catalog/v/07-0398