Learn More
Strem, An Ascensus Company CAS# 67-43-6. 25g. Diethylenetriaminepentaacetic acid, 99% DTPA. MFCD00004289
SDP
Catalog No. 5081468
Encompass
CAS# 67-43-6. 25g. Diethylenetriaminepentaacetic acid, 99% DTPA. MFCD00004289. Molecular Weight: 393.35. Molecular Formula: (HO2CCH2)2N(CH2)2N(CH2CO2H)(CH2)2N(CH2CO2H)2. Color/form: white xtl. Strem# 07-0412. http://www.strem.com/catalog/v/07-0412/
Catalog No. 50-814-68 Supplier Strem, An Ascensus Company Supplier No. 07041225G
CAS# 67-43-6. 25g. Diethylenetriaminepentaacetic acid, 99% DTPA. MFCD00004289. Molecular Weight: 393.35. Molecular Formula: (HO2CCH2)2N(CH2)2N(CH2CO2H)(CH2)2N(CH2CO2H)2. Color/form: white xtl. Strem# 07-0412. http://www.strem.com/catalog/v/07-0412/