Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylenetriaminepentaacetic dianhydride

Catalog No. 2840251G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-402-51G 1 g
28-402-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-402-51G Supplier Sigma Aldrich Supplier No. 2840251G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylenetriaminepentaacetic dianhydride

Specifications

CAS 23911-26-4
Melting Point 182°C to 184°C
Molecular Formula C14H19N3O8
Linear Formula C14H19N3O8
Quantity 1 g
MDL Number MFCD00010697
Synonym N,N-Bis[2-(2,6-dioxomorpholino)ethyl]glycine; DTPA anhydride; DTPA dianhydride
Molecular Weight (g/mol) 357.32
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.