Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylmethoxyborane Solution

Catalog No. 328839100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-883-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 32-883-9100ML Supplier Sigma Aldrich Supplier No. 328839100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in: Studying a catalyst for ring-opening metathesis polymerization / vinyl insertion polymerizationReformatsky / quaternary Claisen condensationsEnantioselective synthesis of carba-furanose sugarsBorane-mediated control radical polymerization for synthesis of fluoropolymersHomologation reactions with sulfonium ylidesTrialkylborane promoted adhesion to low surface energy plastics

Specifications

CAS 81790-10-5
Density 1.121 g/mL at 25°C
Boiling Point 82°C to 85°C (60 mmHg, lit.)
Molecular Formula C6H13BrSi
Linear Formula H2C=C(Br)CH2Si(CH3)3
Quantity 100 mL
MDL Number MFCD00043054
Synonym Diethylborinic acid methyl ester; Methyl diethylborinate
Molecular Weight (g/mol) 193.16 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.