Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylmethylvinylsilane, 98+%, Thermo Scientific™

Catalog No. AAB2321406
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2321406 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2321406 Supplier Thermo Scientific Chemicals Supplier No. B2321406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18292-29-0
Quantity 5g
Molecular Formula C7H16Si
Linear Formula (CH3CH2)2CH3SiCH=CH2
MDL Number MFCD00041344
UN Number UN1993
Flash Point 1°C (35°F)
Synonym diethyl methyl vinyl silane, diethylmethylvinylsilane, diethyl-methyl-vinyl-silane, ethenyldiethylmethylsilane, silane, diethylmethylvinyl, silane, ethenyldiethylmethyl, vinyldiethylmethylsilane, methyldiethylvinylsilane, diethyl-methylvinylsilane
InChI Key BAVNDESSHRPRRF-UHFFFAOYSA-N
SMILES CC[Si](C)(CC)C=C
IUPAC Name ethenyldiethylmethylsilane
Molecular Weight (g/mol) 128.29
PubChem CID 29020
Formula Weight 128.29
Percent Purity ≥98%
Refractive Index 1.422
Density 0.75
Chemical Name or Material Diethylmethylvinylsilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P280a-P240-P241-P303+P361+P353-P501a
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.