Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylphenylphosphine 97.0+%, TCI America™
SDP

Catalog No. D10195ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10195ML 5 mL
D10191ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10195ML Supplier TCI America Supplier No. D10195ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1605-53-4
Molecular Formula C10H15P
Molecular Weight (g/mol) 166.20
MDL Number MFCD00015172
InChI Key LVTCZSBUROAWTE-UHFFFAOYSA-N
Synonym diethylphenylphosphine, diethylphenylphosphene, diethyl phenyl phosphane, phosphine, diethylphenyl, diethyl phenyl phosphine, phenyldiethylphosphine, diethylphenyl phosphine, acmc-209dl7
PubChem CID 74149
IUPAC Name diethyl(phenyl)phosphane
SMILES CCP(CC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 5 mL
Formula Weight 166.20
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylphenylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.