Learn More
Strem, An Ascensus Company CAS# 627-49-6. 5g. Diethylphosphine, 99%. MFCD00015169
SDP
Supplier:  Strem, An Ascensus Company 1512105G
Encompass
CAS# 627-49-6. 5g. Diethylphosphine, 99%. MFCD00015169. Molecular Weight: 90.11. Molecular Formula: (C2H5)2PH. Color/form: colorless liq. Strem# 15-1210. http://www.strem.com/catalog/v/15-1210/
Catalog No. 50-204-5241