Learn More
Strem, An Ascensus Company CAS# 762-04-9. 100g. Diethylphosphite, min. 95%. MFCD00044573
SDP
Catalog No. 5090114203
Encompass
CAS# 762-04-9. 100g. Diethylphosphite, min. 95%. MFCD00044573. Molecular Weight: 138.11. Molecular Formula: (C2H5O)2P(O)H. Color/form: colorless liq. Strem# 93-1521. http://www.strem.com/catalog/v/93-1521/
Catalog No. 50-901-14203 Supplier Strem, An Ascensus Company Supplier No. 931521100G
CAS# 762-04-9. 100g. Diethylphosphite, min. 95%. MFCD00044573. Molecular Weight: 138.11. Molecular Formula: (C2H5O)2P(O)H. Color/form: colorless liq. Strem# 93-1521. http://www.strem.com/catalog/v/93-1521/