Learn More
Gold Biotechnology Inc Diethylpyrocarbonate (DEPC) 25 mL
SDP
Catalog No. NC2942872
Encompass_Preferred
Diethylpyrocarbonate (DEPC) is a moisture sensitive product that will readily decompose to ethanol and carbon dioxide in aqueous solution. DEPC can inhibit ryanodine binding to the ryanodine/Ca2+ receptor channel in a dose-and time-dependent manner, or work as a nuclease inhibitor. DEPC is often used to treat water used in the handling of RNA to remove RNases and reduce the risk of the RNA being degraded.
Catalog No. NC2942872 Supplier Gold Biotechnology Inc Supplier No. D34025
Diethylpyrocarbonate (DEPC) is a moisture sensitive product that will readily decompose to ethanol and carbon dioxide in aqueous solution. DEPC can inhibit ryanodine binding to the ryanodine/Ca2+ receptor channel in a dose-and time-dependent manner, or work as a nuclease inhibitor. DEPC is often used to treat water used in the handling of RNA to remove RNases and reduce the risk of the RNA being degraded.