Learn More
Apexbio Technology LLC Diethylstilbestrol 56-53-1 200mg
SDP
Catalog No. 502240760
Encompass_Preferred
Diethylstilbestrol (56-53-1) is a small-molecule inhibitor targeting 11 -hydroxysteroid dehydrogenase type 2 (HSD11B2) and interacting with estrogen receptors It is designed to modulate hormonal signaling and cellular steroid metabolism thereby regulating processes such as apoptosis and autophagy Diethylstilbestrol exerts its biological activity primarily through the inhibition of HSD11B2 and modulation of estrogen receptor-mediated signaling In cell-based studies Diethylstilbestrol induces oxidative DNA damage and triggers apoptotic responses in spermatogonial stem cells as well as promotes autophagy in thymic cells Based on these pharmacological properties Diethylstilbestrol holds research potential in exploring estrogen receptor-mediated signaling hormone-regulated molecular events apoptosis pathways and autophagy mechanisms in vitro
This item is not returnable. View return policy
Catalog No. 50-224-0760 Supplier Apexbio Technology LLC Supplier No. B2088200
Diethylstilbestrol (56-53-1) is a small-molecule inhibitor targeting 11 -hydroxysteroid dehydrogenase type 2 (HSD11B2) and interacting with estrogen receptors It is designed to modulate hormonal signaling and cellular steroid metabolism thereby regulating processes such as apoptosis and autophagy Diethylstilbestrol exerts its biological activity primarily through the inhibition of HSD11B2 and modulation of estrogen receptor-mediated signaling In cell-based studies Diethylstilbestrol induces oxidative DNA damage and triggers apoptotic responses in spermatogonial stem cells as well as promotes autophagy in thymic cells Based on these pharmacological properties Diethylstilbestrol holds research potential in exploring estrogen receptor-mediated signaling hormone-regulated molecular events apoptosis pathways and autophagy mechanisms in vitro