Learn More
Medchemexpress LLC Diethylstilbestrol-d3 | 58322-36-4 | ≥98% | C18H17D3O2 | 25MG
SDP
Catalog No. 5000356758
Encompass_Preferred
Diethylstilbestrol-d3 is deuterium labeled Diethylstilbestrol. It is for research use only and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-56758 Supplier Medchemexpress LLC Supplier No. 25MGHY14598S
Diethylstilbestrol-d3 is deuterium labeled Diethylstilbestrol. It is for research use only and can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.