Learn More
Apexbio Technology LLC DIETHYLSTILBESTROL-D4-10MG
SDP
Catalog No. 5000570233
Encompass_Preferred
Diethylstilbestrol-d4 (CAS No 56-53-1) is a deuterium-labeled synthetic estrogen used primarily in biomedical research Originating as a derivative of diethylstilbestrol its mechanism of action involves binding to estrogen receptors influencing gene expression and cellular pathways related to growth and differentiation In vitro it exhibits activity typically in the nanomolar to low micromolar range This compound is frequently employed in cell models and animal studies specifically for endocrine and hormonal research It serves as a valuable tool in drug screening particularly for evaluating estrogenic and anti-estrogenic effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70233 Supplier Apexbio Technology LLC Supplier No. SL102210
Diethylstilbestrol-d4 (CAS No 56-53-1) is a deuterium-labeled synthetic estrogen used primarily in biomedical research Originating as a derivative of diethylstilbestrol its mechanism of action involves binding to estrogen receptors influencing gene expression and cellular pathways related to growth and differentiation In vitro it exhibits activity typically in the nanomolar to low micromolar range This compound is frequently employed in cell models and animal studies specifically for endocrine and hormonal research It serves as a valuable tool in drug screening particularly for evaluating estrogenic and anti-estrogenic effects Experimental concentrations typically depend on the specific research design