Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethylstilbestrol-d8 | 91318-10-4 | MFCD01074086 | 99.7% | C18H12D8O2 | 10MG
SDP

Catalog No. 5000258441
Encompass_Preferred

Diethylstilbestrol-d8 is the deuterium-labeled analogue of diethylstilbestrol supplied as a high-purity powder for research and analytical use. It is intended as a stable isotope internal standard for LC-MS, pharmacokinetic studies, and quantitative assays where mass differentiation from the parent compound is required.

  • High isotopic purity suitable for mass spectrometry internal standard.
  • Purity 99.7% as supplied.
  • Molecular formula C18H12D8O2 and molecular weight 276.40.
  • Recommended solvent: DMSO, 50 mg/mL with ultrasonic and warming.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Supplied in small research pack sizes appropriate for analytical workflows.

Catalog No. 50-002-58441 Supplier Medchemexpress LLC Supplier No. HY14598S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethylstilbestrol-d8 is the deuterium-labeled analogue of diethylstilbestrol supplied as a high-purity powder for research and analytical use. It is intended as a stable isotope internal standard for LC-MS, pharmacokinetic studies, and quantitative assays where mass differentiation from the parent compound is required.

  • High isotopic purity suitable for mass spectrometry internal standard.
  • Purity 99.7% as supplied.
  • Molecular formula C18H12D8O2 and molecular weight 276.40.
  • Recommended solvent: DMSO, 50 mg/mL with ultrasonic and warming.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Supplied in small research pack sizes appropriate for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.