Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylstilbestrol dipropionate, MilliporeSigma™ Supelco™

Catalog No. 111012923 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2923 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2923 Supplier MilliporeSigma Supelco Supplier No. D38811G

Diethylstilbestrol dipropionate is a potent synthetic form of estrogen, belonging to the class of xenobiotic anabolic compounds commonly used for the fattening of cattle.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 130-80-3
Physical Form Neat
Linear Formula C24H28O4
MDL Number MFCD00027002
Synonym (E)-3,4-Diphenyl-3-hexene p,p′-dipropionate; Stilbestrol dipropionate
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 g
Formula Weight 380.48
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.