Learn More
Medchemexpress LLC Diethyltoluamide-d10 | 1215576-01-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0978S1MG
Encompass_Preferred
Diethyltoluamide-d10 is the deuterium labeled Diethyltoluamide, the most common active ingredient in insect repellents. It provides protection against mosquitoes, ticks, fleas, chiggers, leeches, and many other biting insects.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08877