Learn More
Medchemexpress LLC Diethyltoluamide-d7 | 1219799-37-7 | 98.5% | 1 MG
SDP
Catalog No. 5000408835
Encompass_Preferred
Diethyltoluamide-d7 is the deuterium labeled Diethyltoluamide. It is the most common active ingredient in insect repellents, providing protection against mosquitoes, ticks, fleas, chiggers, leeches, and many other biting insects. This product is for research use only.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08835 Supplier Medchemexpress LLC Supplier No. HYB0978S11MG
Diethyltoluamide-d7 is the deuterium labeled Diethyltoluamide. It is the most common active ingredient in insect repellents, providing protection against mosquitoes, ticks, fleas, chiggers, leeches, and many other biting insects. This product is for research use only.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS