Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyltoluamide-d7 | 1219799-37-7 | 98.5% | 198.31 | C12H10D7NO | 10 MG
SDP

Catalog No. 5000297731
Encompass_Preferred

Diethyltoluamide-d7 is a deuterium-labeled analogue of N,N-diethyl-m-toluamide (DEET) used as an isotope-labeled reference standard for analytical chemistry. It is intended for use as an internal standard in mass spectrometry, calibration, and method validation, supplied in milligram quantities with high isotopic enrichment and purity.

  • Deuterium-labeled internal standard (D7) for mass spectrometry.
  • High purity (98.5%) suitable for quantitative analysis.
  • Molecular weight 198.31 g/mol.
  • CAS number 1219799-37-7 for unambiguous identification.
  • Supplied in small milligram quantities for analytical workflows.

Catalog No. 50-002-97731 Supplier Medchemexpress LLC Supplier No. HYB0978S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyltoluamide-d7 is a deuterium-labeled analogue of N,N-diethyl-m-toluamide (DEET) used as an isotope-labeled reference standard for analytical chemistry. It is intended for use as an internal standard in mass spectrometry, calibration, and method validation, supplied in milligram quantities with high isotopic enrichment and purity.

  • Deuterium-labeled internal standard (D7) for mass spectrometry.
  • High purity (98.5%) suitable for quantitative analysis.
  • Molecular weight 198.31 g/mol.
  • CAS number 1219799-37-7 for unambiguous identification.
  • Supplied in small milligram quantities for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.