Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylzinc

Catalog No. 502906971
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-6971 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-6971 Supplier Sigma Aldrich Supplier No. 66872925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylzinc
TRUSTED_SUSTAINABILITY

Specifications

CAS 557-20-0
Melting Point -28°C
Density 1.205 g/mL (at 25°C)
Boiling Point 117°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2 Zn
Refractive Index n20/D 1.498 (literature)
Quantity 25 g
MDL Number MFCD00009021
Synonym DEZn; Et2Zn; DEZ; Zincdiethyl
Molecular Weight (g/mol) 123.51
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.