Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylzinc Solution

Catalog No. 406023100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-602-3100ML 100 mL
40-602-3800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 40-602-3100ML Supplier Sigma Aldrich Supplier No. 406023100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylzinc solution can be used in the synthesis of:Bis(pyridylpyrrolyl)zinc luminescent complexes.A versatile building block, 5-(ketoaryl)thiazole.ZnxCd1-xSe nanocrystals having high luminescence properties.

Specifications

CAS 557-20-0
Density 0.74 g/mL at 25°C
Boiling Point 98°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2Zn
MDL Number MFCD00009021
Quantity 100 mL
Synonym Zincdiethyl
Molecular Weight (g/mol) 123.51 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.