Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylzinc Solution

Catalog No. 296112100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-611-2100ML 100 mL
29-611-22L 2 L
29-611-2800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 29-611-2100ML Supplier Sigma Aldrich Supplier No. 296112100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylzinc solution can be used as a reagent to prepare:+a-trifluoromethyl ketones from +a, +a-unsaturated ketones with Rh (I) catalyst.Alkynylzinc reagents which are used in cross-coupling reactions.Chiral secondary alcohols from aromatic and aliphatic aldehydes in presence of camphor-derived +a-amino alcohol.1, 4 conjugate addition product with enones.+a-fluoroacrylates from aldehydes and ketones.

Specifications

CAS 557-20-0
Density 0.726 g/mL at 25°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2Zn
MDL Number MFCD00009021
Quantity 100 mL
Synonym Zincdiethyl
Molecular Weight (g/mol) 123.51 g/mol
Biological Activity Central Nervous System, Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.