Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylzinc Solution

Catalog No. 220809800G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
800 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-080-9800G 800 g
22-080-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-080-9800G Supplier Sigma Aldrich Supplier No. 220809800G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed together with CF3I in a Rh(I) catalyzed preparation of +a-trifluoromethyl ketones from +a,+â-unsaturated ketones. Promoter for the Wittig reaction of carbonyl compounds with dibromofluoroacetate and triphenylphosphine leading to +a-fluroroacrylates. Ni catalyzed coupling with cyclic anhydrides.

Specifications

CAS 557-20-0
Density 0.915 g/mL at 25°C
Molecular Formula C4H10Zn
Linear Formula (C2H5)2Zn
MDL Number MFCD00009021
Quantity 800 g
Synonym Zincdiethyl
Molecular Weight (g/mol) 123.51 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.