Learn More
Stoelting™ Dietrich Bulldog Clamp
SDP
Catalog No. 10000480 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-480 Curved
10-000-479 Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-480 Supplier Stoelting™ Supplier No. 5212039P
Add to Cart
Add to Cart
Stoelting™ Dietrich Bulldog Clamp is useful in surgeries that require precise blood stoppage.
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Specifications
For Use With (Application) Neuroscience
Length (English) 2 in.
Model Curved