Learn More
Fine Science Tools Dietrich Bulldog Clamps, Stainless Steel, 50 mm
SDP
Catalog No. NC9971994
Encompass
Product: Dietrich Bulldog Clamp - Angled/50mm/ Shape: Angled/ Jaw Dimensions: 8.6 x 1.3mm/ Clamping Pressure: Light/ Gram Pressure: 50g/ Alloy: Stainless Steel/ Length: 50mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9971994 Supplier Fine Science Tools Supplier No. 1803945
Product: Dietrich Bulldog Clamp - Angled/50mm/ Shape: Angled/ Jaw Dimensions: 8.6 x 1.3mm/ Clamping Pressure: Light/ Gram Pressure: 50g/ Alloy: Stainless Steel/ Length: 50mm/ Serrated: Yes/ Unit: Each